ZRB2593
Anti-TRF1/TERF1 Antibody, clone 6I14 ZooMAb® Rabbit Monoclonal
recombinant, expressed in HEK 293 cells
Synonym(s):
NIMA-interacting protein 2, TTAGGG repeat-binding factor 1, Telomeric protein Pin2/TRF, Telomeric repeat-binding factor 1
About This Item
Recommended Products
biological source
rabbit
Quality Level
recombinant
expressed in HEK 293 cells
conjugate
unconjugated
antibody form
purified antibody
antibody product type
primary antibodies
clone
recombinant monoclonal
description
clone 6I14
recombinant, expressed in HEK 293 cells
product line
ZooMAb® learn more
form
lyophilized
mol wt
calculated mol wt 50.25 kDa
observed mol wt ~55 kDa
purified by
using Protein A
species reactivity
human
packaging
antibody small pack of 25
greener alternative product characteristics
Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
enhanced validation
recombinant expression
Learn more about Antibody Enhanced Validation
sustainability
Greener Alternative Product
technique(s)
affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable
isotype
IgG
epitope sequence
N-terminal
Protein ID accession no.
UniProt accession no.
shipped in
ambient
storage temp.
2-8°C
Gene Information
human ... TERF1(7013)
Related Categories
General description
Specificity
Immunogen
Application
Evaluated by Western Blotting in Raji cell nuclear extract.
Western Blotting Analysis (WB): A 1:10,000 dilution of this antibody detected TRF1/TERF1 in Raji cell nuclear extract.
Tested Applications
Western Blotting Analysis: A 1:10,000 dilution from a representative lot detected TRF1/TERF1 in HeLa cell nuclear extract.
Affinity Binding Assay: A representative lot of this antibody bound TRF1/TERF1 peptide with a KD of 2.8 x 10-6 in an affinity binding assay.
Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected TRF1/TERF1 in Human testis tissue sections.
Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected TRF1/TERF1 in Raji cells.
Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.
Target description
Physical form
Reconstitution
Storage and Stability
Legal Information
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service