Skip to Content
Merck
All Photos(13)

Key Documents

ZRB2244

Sigma-Aldrich

Anti-OXCT1 Antibody, clone 1B11 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

3-oxoacid CoA-transferase 1, EC:2.8.3.5, SCOT, SCOT-s, Somatic-type succinyl-CoA:3-oxoacid CoA-transferase, Succinyl-CoA:3-ketoacid coenzyme A transferase 1 mitochondrial, Succinyl-CoA:3-oxoacid CoA transferase

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
€187.00
4 X 25 μL
€385.00

€187.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
€187.00
4 X 25 μL
€385.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€187.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1B11, recombinant monoclonal

description

1B11 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 56.16 kDa
observed mol wt ~Target molecular weight 52 kDa

purified by

using Protein A

species reactivity

rat, mouse, human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

Internal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... OXCT1(5019)

Related Categories

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry. This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1B11 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects 3-oxoacid CoA-transferase 1(OXCT1). It target an epitope within 16 amino acids from the internal region.

Immunogen

KLH-conjugated linear peptide corresponding to 16 amino acids from the internal region of human 3-oxoacid CoA-transferase 1(OXCT1).

Application

Quality Control Testing

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected OXCT1 in HeLa cell lysate.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected OXCT1 in lysates from A431 cells, Human heart and Rat heart tissue, and NIH3T3 cells.

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected OXCT1 in Human heart tissue sections.

Affinity Binding Assay: A representative lot of this antibody bound OXCT1 peptide with a KD of 5.7 x 10-6 in an affinity binding assay.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected OXCT1 in HeLa cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Succinyl-CoA:3-ketoacid coenzyme A transferase 1, mitochondrial (UniProt: P55809; also known as EC:2.8.3.5, SCOT, 3-oxoacid CoA-transferase 1, Somatic-type succinyl-CoA:3-oxoacid CoA-transferase, SCOT-s, Succinyl-CoA:3-oxoacid CoA transferase) is encoded by the OXCT1 (also known as OXCT, SCOT) gene (Gene ID: 5019) in human.  OXCT1 is a homodimeric, mitochondrial enzyme of the 3-oxoacid CoA-transferase family. It catalyzes the transfer of the CoA moiety from succinyl-CoA to acetoacetate, producing acetoacetyl-CoA. Acetoacetyl-CoA is further metabolized by acetoacetyl-CoA thiolase into two acetyl-CoA molecules, which enter the citric acid cycle for energy production.  The catalysis by OXCT1 proceeds via a classical ping-pong mechanism where each monomer binds one molecule of the first substrate succinyl-CoA, at the interface between the two domains where it is attacked by the strictly conserved Glu344 to form an enzyme-CoA thioester intermediate. This intermediate is then attacked by the second incoming substrate acetoacetate to yield the final product acetoacetyl-CoA. Mutations in OXCT1 gene are known to cause succinyl-CoA:3-oxoacid CoA transferase deficiency (SCOTD), a rare autosomal disorder that leads to episodic ketoacidosis. OXCT1 is abundantly expressed in extrahepatic tissues, including heart, brain, kidney, skeletal muscle, and lung. Two isoforms of OXCT1 have been described that are produced by alternative splicing.  This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Grünert, SC., et al. (2021). Biochimie. 183; 55-62; Zheng, DJ., et al. (2018). J. Pediatr. Intensive Care. 7(1); 62-66).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service