Skip to Content
Merck
All Photos(11)

Key Documents

ZRB2063

Sigma-Aldrich

Anti-PDIA4 Antibody, clone 1B20 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Protein disulfide-isomerase A4;EC:5.3.4.1;Endoplasmic reticulum resident protein 70;ER protein 70;ERp70;Endoplasmic reticulum resident protein 72;ER protein 72;ERp-72;ERp72

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
€187.00
4 X 25 μL
€385.00

€187.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
€187.00
4 X 25 μL
€385.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€187.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1B20, recombinant monoclonal

description

1B20 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 72.93 kDa
observed mol wt ~Target molecular weight 73 kDa

purified by

using Protein A

species reactivity

rat, mouse, human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal half

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... PDIA4(9601)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1B20 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects Protein disulfide-isomerase A4 (PDIA4). It targets an epitope within 22 amino acids from the N-terminal half.

Immunogen

KLH-conjugated linear peptide corresponding to 22 amino acids from the N-terminal half of human Protein disulfide-isomerase A4 (PDIA4).

Application

Quality Control Testing

Evaluated by Western Blotting in A549 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected PDIA4 in A549 cell lysate.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected PDIA4 in NIH3T3, Panc-1, and PC12 cell lysates.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected PDIA4 in NIH3T3 cells.

Affinity Binding Assay: A representative lot of this antibody bound PDIA4 peptide with a KD of 3.3 x 10-7 in an affinity binding assay.

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected PDIA4 in human liver tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Protein disulfide-isomerase A4 (UniProt: P13667; also known as EC:5.3.4.1, Endoplasmic reticulum resident protein 70, ER protein 70, ERp70, Endoplasmic reticulum resident protein 72, ER protein 72, ERp-72, ERp72) is encoded by the PDIA4 (also known as ERP70, ERP72) gene (Gene ID: 9601) in human. Protein disulfide isomerases (PDIs) are a family of multifunctional enzymes with oxidoreductase and chaperone activities that regulate the redox state of proteins and maintain their proper folding and function. The PDI family has 21 members and PDIA4 is the biggest of the PDIs with three CGHC motifs, two in the N-terminal and one in the C-terminal half. It is a part of a large chaperone multiprotein complex. It is synthesized with a signal peptide (aa 1-20), which is subsequently cleaved off to produce the mature protein that mainly resides in the lumen of endoplasmic reticulum (ER) where the KEEL sequence at its C-terminus prevents its secretion from ER. PDIA4 is implicated in the growth and death of tumor cells and its expression is shown to be upregulated in a variety of tumor cell lines and in human lung adenocarcinoma tissues. Its levels are also upregulated during ER stress. It is reported to promote lung cancer development via regulation of STAT3/VEGF axis in T and B lymphocytes of cancer stroma. It is reported to regulate the activation and degradation of procaspases 3 and 7 by caspase 8. It controls the stability and function of these caspases in a CGHC motif-dependent manner. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Chen, TY., et al. (2022). Clin. Transl. Med. 12(2); e606; Kuo, T-F., et al. (2017). Oncogene. 36(39); 5484-5496).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

0.3 mg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service