Skip to Content
Merck
All Photos(15)

Key Documents

ZRB1938

Sigma-Aldrich

Anti-Cytokeratin-20 Antibody, clone 1E20 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Keratin, type I cytoskeletal 20;CK-20;Keratin-20;K20;Protein IT

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
€191.00
4 X 25 μL
€393.00

€191.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
€191.00
4 X 25 μL
€393.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

€191.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1E20, recombinant monoclonal

description

1E20 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 48.49 kDa
observed mol wt ~48 kDa

purified by

using Protein A

species reactivity

human

species reactivity (predicted by homology)

monkey

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... KRT20(54474)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1E20 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects Cytokeratin-20. It targets an epitope within 23 amino acids from the C-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 23 amino acids from the C-terminal region of human Cytokeratin-20.

Application

Quality Control Testing
Evaluated by Western Blotting in HT-29 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Cytokeratin-20 in HT-29 cell lysate.
Tested applications
Flow Cytometry Analysis: 0.1 μg from a representative lot detected Cytokeratin-20 in one million HT-29 cells.
Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected Cytokeratin-20 in human small intestine tissue sections.
Affinity Binding Assay: A representative lot of this antibody bound Cytokeratin-20 peptide with a KD of 6.8 x 10-8 in an affinity binding assay.
Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected Cytokeratin-20 in HT-29 cells.
Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Evaluated by Western Blotting in HT-29 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Cytokeratin-20 in HT-29 cell lysate.

Target description

Keratin, type I cytoskeletal 20 (UniProt: P35900; also known as Cytokeratin-20, CK-20, Keratin-20, K20, Protein IT) is encoded by the KRT20 gene (Gene ID: 54474) in human. Cytokeratin-20 is a heterotetramer of two type I and two type II keratins of the intermediate filament family. It is expressed predominantly in the intestinal epithelium and in luminal cells of colonic mucosa. Its expression is also observed in the Merkel cells of keratinized oral mucosa. It is recognized as a marker of intestinal cell differentiation and is also an excellent marker for certain types of carcinomas, such as adenocarcinomas of the colon, transitional cell carcinomas of the bladder and Merkel cell tumors of the skin. Cytokeratin-20 It plays a significant role in maintaining keratin filament organization in intestinal epithelia and when phosphorylated, it also plays a role in the secretion of mucin in the small intestine. It is reported to undergo hyperphosphorylation at serine 13 during the early stages of apoptosis, which becomes less prominent during the later stages. Phosphorylation at serine 13 is shown to increase in response to stress brought on by cell injury. During apoptosis it is cleaved at Asp 228 by caspases. This ZooMAbZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Moll, R., et al. (1992). Am. J. Pathol. 140(2); 427-447).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service