Skip to Content
Merck
All Photos(12)

Documents

ZRB1830

Sigma-Aldrich

Anti-NOTCH2 Antibody, clone 1B14-5 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1B14, monoclonal
recombinant monoclonal

description

1B14 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt ~265.41 kDa
observed mol wt ~110 kDa

species reactivity

human, mouse, rat

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1B14-5 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects NOTCH2. It targets an epitope within 18 amino acids from the C-terminal, cytoplasmic domain.

Immunogen

KLH-conjugated linear peptide corresponding to 18 amino acids from the C-terminal, cytoplasmic domain of human NOTCH2.

Application

Quality Control Testing

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected NOTCH2 in HeLa cell lysate.

Tested Applications

Western Blotting Analysis: A 1:10,000 dilution from a representative lot detected NOTCH2 in NIH3T3 and C6 cell lysates.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected NOTCH2 in human kidney tissue sections.

Affinity Binding Assay: A representative lot of this antibody bound Notch2 peptide with a KD of 1.3 x 10-6 in an affinity binding assay.

Immunocytochemistry Analysis: A 1:1,000 dilution from a representative lot detected NOTCH2 in MCF7 cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-NOTCH2, clone 1B14-5 ZooMAb®, Cat. No. ZRB1830, is a recombinant Rabbit monoclonal antibody that specifically targets NOTCH2 and is tested for use in Affinity Binding Assay, Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.

Target description

Neurogenic locus notch homolog protein 2 (UniProt: Q04721; also known as Notch 2, hN2, NOTCH2) is encoded by the NOTCH2 gene (Gene ID: 4853) in human. Notch is a single-pass type I membrane protein that is synthesized with a signal peptide (aa 1-25), which is subsequently cleaved off to generate the mature form that contains a long extracellular domain (aa 26-2677), a transmembrane domain (aa 1678-1698), and a cytoplasmic domain (aa 1699-2471). It is evolutionarily conserved and is implicated in cell-fate determination, tissue patterning, morphogenesis, and cell differentiation. It is expressed in the brain, heart, kidney, lung, skeletal muscle and liver and is ubiquitously expressed in the embryo. Notch is synthesized in the endoplasmic reticulum as an inactive form which is proteolytically cleaved by a furin-like convertase in the trans-Golgi network before it reaches the plasma membrane to yield an active, ligand-accessible form. It serves as a receptor for Jagged-1, Jagged-2, and Delta-1 to regulate cell-fate determination. Following ligand binding, Notch is cleaved by TNF-alpha converting enzyme (S2 cleavage) to yield a membrane-associated intermediate fragment called notch extracellular truncation, which is then cleaved by g-secretase (S3 cleavage) to release a notch-derived peptide containing the intracellular domain (NICD; aa 1697-2471) from the membrane that translocates to the nucleus and initiates transcription events by binding the DNA binding protein CSL. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Yin, L., et al (2010). Biochem. Pharmacol. 80(5); 690-701).

Physical form

Purified recombinant mouse monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service