Skip to Content
Merck
All Photos(26)

Documents

ZRB1105

Sigma-Aldrich

Anti-C-EBP beta Antibody, clone 8G18, ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit (recombinant)

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

8G18, recombinant monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 36.11 kDa
observed mol wt ~42 kDa

species reactivity

human, mouse

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable (peptide)

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

mouse ... Cebpb(12608)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 8G18 ZooMAb is a rabbit recombinant monoclonal antibody that specifically detects human CCAAT/enhancer-binding protein beta (C/EBP beta). It targets an epitope with in the N-terminal region.

Immunogen

His-tagged recombinant fragment corresponding to 150 amino acids from the N-terminal half of human CCAAT/enhancer-binding protein beta (C/EBP beta).

Application

Anti-C-EBP beta, clone 8G18 ZooMAb , Cat. No. ZRB1105, is a highly specific rabbit recombinant monoclonal antibody that targets C-EBP beta and has been tested for use in Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.
Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected C-EBP beta in HeLa, A431, HUVEC, NIH 3T3 and HepG2 cells.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected C-EBP beta in human colon and mouse colon tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

CCAAT/enhancer-binding protein beta (UniProt: P17676; also known as C/EBP beta, Liver activator protein, LAP, Liver-enriched inhibitory protein, LIP, Nuclear factor NF-IL6, Transcription factor 5, TCF-5) is encoded by the CEBPB (also known as TCF5) gene (Gene ID: 1051) in human. C/EBP beta is a highly conserved B-Zip family member that acts as a transcription factor regulating the expression of genes involved in immune and inflammatory responses. It is expressed at low levels in the lung, kidney and spleen and is induced by endoplasmic reticulum stress. It can bind to DNA either as a homodimer or a heterodimer with ATF4. C/EBP beta plays a significant role in adipogenesis, as well as in the gluconeogenesis, liver regeneration, and hematopoiesis. During adipogenesis, it is rapidly expressed and, after activation by phosphorylation, induces CEBPA and PPARG, which turn on the series of adipocyte genes giving rise to the adipocyte phenotype. It has a pro-mitotic effect on many cell types, including hepatocytes and adipocytes, but has an anti-proliferative effect on T-cells by repressing MYC expression, facilitating differentiation along the T-helper 2 lineage. C/EBP beta is phosphorylated at threonine 235 by MAPK and CDK2 that serves to prime phosphorylation at threonine 226 and serine 231 by GSK3beta and acquire DNA-binding as well as transactivation activities that are required to induce adipogenesis. However, O-glycosylation at serine 227 and serine 228 is shown to prevent phosphorylation on threonine 235, serine 231, and threonine 226 and reduces its DNA binding activity that delays adipocyte differentiation. This ZooMAb recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose. Normal appearance is a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service