Skip to Content
Merck
All Photos(14)

Key Documents

ZRB1076

Sigma-Aldrich

Anti-PER2 Antibody, clone 1G17 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Circadian clock protein PERIOD 2, Period circadian protein homolog 2, hPER2

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
€162.00
4 X 25 μL
€334.00

€162.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
€162.00
4 X 25 μL
€334.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€162.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1G17, recombinant monoclonal
monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 136.58 kDa

species reactivity

human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... PER2(8864)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 1G17 is a ZooMAb rabbit recombinant monoclonal antibody that specifically detects Period circadian protein homolog 2 (PER2). It targets an epitope within 21 amino acids from the N-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 21 amino acids from the N-terminal region of human Period circadian protein homolog 2 (PER2).

Application

Anti-PER2, clone 1G17 ZooMAb, Cat. No. ZRB1076, is a recombinant Rabbit monoclonal antibody that specifically targets PER2 and has been tested for use in Affinity Binding Assay, Immunocytochemistry, and Immunohistochemistry (Paraffin).
Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected PER2 in human retina tissue sections.

Immunocytochemistry Analysis: A 1:1000 dilution from a representative lot detected PER2 in HeLa cells.

Affinity Binding Assay: A representative lot of this antibody bound PER2 peptide with a KD of 7.2 x 10-8 in an affinity binding assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

Period circadian protein homolog 2 (UniProt: O15055; also known as hPER2, Circadian clock protein PERIOD 2, PER2) is encoded by the PER2 (also known as KIAA0347) gene (Gene ID: 8864) in human. PER2 is a homodimeric protein that is widely expressed and is high levels are found in skeletal muscle and pancreas. It serves as a transcriptional repressor and is a core component of the circadian clock along with other clock components (CLOCK, NPAS2, ARNTL/BMAL1, ARNTL2/BMAL2, PER1, PER2, PER3, CRY1 and CRY2) it plays a critical role in rhythm generation. Its levels oscillate diurnally, and its rhythmic levels are critical for the generation of circadian rhythms in central as well as peripheral clocks. Transcription factors, CLOCK or NPAS2 and ARNTL/BMAL1 or ARNTL2/BMAL2, form the positive limb of the feedback loop, act in the form of a heterodimer and activate the transcription of core clock genes and clock-controlled genes. The core clock genes: PER1/2/3 and CRY1/2 which are transcriptional repressors form the negative limb of the feedback loop and interact with the CLOCK/NPAS2, ARNTL/BMAL1, ARNTL2/BMAL2 heterodimer inhibiting its activity. PER2 shuttles between the nucleus and cytoplasm and this shuttling is affected by interaction with other circadian core oscillator proteins and/or by phosphorylation. It translocates to the nucleus after phosphorylation by CSNK1D or CSNK1E. PER2 contains a nuclear localization signal (aa 789-805) and three nuclear export signals (aa 111-120, 462-471, and 989-996). Mutations in PER2 gene have been linked to advanced sleep phase syndrome. This ZooMAb recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service