Skip to Content
Merck
All Photos(7)

Key Documents

ZMS1119

Sigma-Aldrich

Anti-SV40 T Antigen Antibody, clone PAb416 ZooMAb® Mouse Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Anti-XLF/NHEJ1 clone 1C16 ZooMAb® Rabbit Monoclonal, LT, LT-AG, Large T antigen

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
€188.00
4 X 25 μL
€386.00

€188.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
€188.00
4 X 25 μL
€386.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

€188.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

PAb416, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 81.62 kDa
observed mol wt ~80 kDa

purified by

using protein G

species reactivity

virus

species reactivity (predicted by homology)

all

packaging

antibody small pack of 25

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgG2aκ

epitope sequence

N-terminal half

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

vaccinia virus ... SV40gp6(29031019)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone PAb416 is a ZooMAb® mouse recombinant monoclonal antibody that specifically detects Large T antigen of Simian virus 40.

Immunogen

Purified large T-antigen of SV40

Application

Quality Control Testing

Evaluated by Western Blotting in lysate from SV40 infected COS-1 cells.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Large T antigen in lysate from SV40 infected COS-1 cells.

Tested Applications

Immunocytochemistry Analysis: A 1:1,000 dilution from a representative lot detected SV40 Large T Antigen in COS-7 cells.

Flow Cytometry Analysis: 0.1 µg from a representative lot detected SV40 Large T Antigen in one million COS-7 Cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Large T antigen (UniProt: P03070; also known as LT, LT-AG) is encoded by the SV40gp6 gene (Gene ID: 29031019) in Simian virus 40. Large T antigen is a key early protein that is reported to be essential for both driving viral replication and inducing cellular transformation. It plays a role in viral genome replication by driving entry of quiescent cells into the cell cycle and by autoregulating the synthesis of viral early mRNA. It displays highly oncogenic activities by corrupting the host cellular checkpoint mechanisms that guard cell division and the transcription, replication, and repair of DNA. It is shown to participates in the modulation of cellular gene expression preceding viral DNA replication and this step involves binding to host key cell cycle regulators retinoblastoma protein RB1/pRb and TP53. It is also known to induce the disassembly of host E2F1 transcription factors from RB1, thus promoting transcriptional activation of E2F1-regulated S-phase genes. It inhibits host TP

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service