Skip to Content
Merck
All Photos(11)

Key Documents

ZMS1091

Sigma-Aldrich

Anti-Heparan Sulfate Antibody, clone T320.11 ZooMAb® Mouse Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

HS;Heparin

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
€191.00
4 X 25 μL
€393.00

€191.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
€191.00
4 X 25 μL
€393.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

€191.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

T320.11, recombinant monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

observed mol wt ~110 kDa

purified by

using protein G

species reactivity

human, rat

species reactivity (predicted by homology)

all

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone T320.11 is a is a ZooMAb® Mouse recombinant monoclonal antibody that detects heparin and heparan sulfate.

Immunogen

Heparan sulfate

Application

Quality Control Testing

Evaluated by Western Blotting in C6 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Heparin/Heparan Sulfate in C6 cell lysate.

Tested applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected Heparin/Heparan Sulfate in U87 and U251 cell lysates.

Immunohistochemistry Applications: A 1:100 dilution from a representative lot detected Heparin/Heparan Sulfate in human colon tissue sections.

Immunofluorescence Analysis: A 1:100 dilution from a representative lot detected Heparin/Heparan Sulfate in frozen human colon tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Evaluated by Western Blotting in C6 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Heparin Sulfate in C6 cell lysate.

Target description

Heparan sulfate (HS) polysaccharides are a family of linear sulfated, heterogeneous polysaccharides found on the cell membrane and in the extracellular matrix as part of heparan sulfate proteoglycans (HSPGs). HSPGs contain a core protein to which heparan sulfate glycosaminoglycan chains are covalently attached. HS contain repeating 1 → 4 linked disaccharide units, in which one monosaccharide is -D-glucosamine residue and the other is a uronic acid. Heparin, a structurally similar polysaccharide, is found within mast cells as a component of serglycin proteoglycans. It is a more sulfated and tissue specific variant of HS. In HS, the d-glucosamine residues are reported to be predominantly N-acetylated, whereas in heparin, they are N-sulfated. Also, in HS the uronates are mainly β-D-glucuronate and in heparin they are predominantly -l-iduronate. Biosynthesis of both HS and heparin begins in the endoplasmic reticulum and proceeds through the Golgi where they are acted upon by several polymerases and modifying enzymes. HS and heparin are known to regulate biological processes through interactions with a large number of proteins. Their poly- and oligosaccharide chains can interact with a very large number of proteins known as heparin binding proteins (HBPs). Changes in HS structure confer the particular protein binding capabilities, which are required for specific biological functions. HS and heparin serve as negatively charged targets for protein binding and attract the positively charged binding sites on proteins via lysine and arginine residues. They can also bind histidine residues under conditions of low pH. Some of these proteins are associated with the extracellular matrix and may be involved in intercellular signaling. This ZooMAbZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Meneghetti, MCZ., et al. (2015). J. R. Soc. Interface. 12(110); 0589; Snigireva, AV., et al. (2015). Cell Adh. Migr. 9(6); 460-468).

Physical form

Purified recombinant mouse monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service