Skip to Content
Merck
All Photos(14)

Key Documents

ZMS1047

Sigma-Aldrich

Anti-dsDNA Antibody, clone AE-2 ZooMAb® Mouse Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

dsDNA

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
€187.00
4 X 25 μL
€385.00

€187.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
€187.00
4 X 25 μL
€385.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€187.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

AE-2, recombinant monoclonal

product line

ZooMAb® learn more

form

lyophilized

purified by

using protein G

species reactivity (predicted by homology)

all

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

dot blot: suitable
immunofluorescence: suitable
immunohistochemistry: suitable

isotype

IgG1

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Related Categories

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry. This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone AE-2 is a ZooMAb® mouse recombinant monoclonal antibody that detects double stranded DNA. May show some cross-reactivity with single-stranded DNA.

Immunogen

Nuclei from Raji Burkitt′s lymphoma Cells.

Application

Quality Control Testing

Evaluated by Immunohistochemistry (Paraffin) in Human tonsil tissue sections.

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution of this antibody detected dsDNA in Human tonsil tissue sections.

Tested Applications

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected dsDNA in human prostate cancer tissue sections.

Dot Blot Analysis: A 1:1,000 dilution from a representative lot detected dsDNA extracted from HeLa cells.

Immunofluorescence Analysis: A 1:100 dilution from a representative lot detected dsDNA in human prostate cancer tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Due to their location in the nucleus and mitochondria, dsDNA is not easily accessible to the immune system. They are quickly degraded by DNases in the cytoplasm and endosomes. However, they can be released from apoptotic cells after exposure to ultraviolet light, infection, and following some drug treatments. The released DNA can then be recognized by anti-DNA antibodies. and compose immune complexes. Extracellular nucleic acids containing immune complexes can be captured by immature dendritic cells (iDCs) via FcγRs. These cells are activated and migrate from peripheral tissues to lymphatic organs, where iDCs undergo maturation.  Anti-dsDNA antibodies are considered to be highly specific markers of Systemic lupus erythematosus (SLE) and autoimmune (lupoid) hepatitis. Their higher levels are observed in about 50 to70% of patients with active, untreated SLE. These antibodies can recognize diverse DNA structures, including single-stranded, Z-DNA, bent or elongated dsDNA, and DNA:RNA hybrids. Anti-dsDNA antibodies may have different subclasses, including IgA, IgE, IgG, and IgM. However, not all of them contribute equally to tissue injuries associated with SLE. IgG and IgA correlate more with disease activity in humans. IgM antibodies seem to be protective by enhancing the elimination of apoptotic material and via immunomodulatory effects. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Wang, X., and Xia, Y. (2019). Front. Immunol. 10; 1667; Haugbro, K., et al. (2004). Ann. Rheum. Dis. 63(4); 386-394).

Physical form

Purified recombinant mouse monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service