Skip to Content
Merck
All Photos(20)

Key Documents

ZMS05637

Sigma-Aldrich

Anti-BMI-1 Antibody, clone F6 ZooMAb® Mouse Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Polycomb complex protein BMI-1, Polycomb group RING finger protein 4, RING finger protein 51

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
€167.00
4 X 25 μL
€345.00

€167.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
€167.00
4 X 25 μL
€345.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€167.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse (recombinant)

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

F6, recombinant monoclonal
monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 36.95 kDa
observed mol wt ~38 kDa

species reactivity

human, mouse

species reactivity (predicted by homology)

rabbit, rat

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG1

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... BMI1(648)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone F6 is a ZooMAb mouse recombinant monoclonal antibody that specifically detects BMI-I. It targets an epitope within the N-terminal region.

Immunogen

Recombinant fragment corresponding to the first 202 amino acids from Mouse Polycomb complex protein BMI-1.

Application

Anti-BMI-1, clone F6 ZooMAb, Cat. No. ZMS05637, is a recombinant mouse monoclonal antibody that specifically targets BMI-1 and is tested for use in Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected BMI-1 in U2OS cell lysate.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected BMI-1 in human placenta tissue sections.

Immunocytochemistry Analysis: A 1:1,000 dilution from a representative lot detected BMI-1 in HeLa, A431, HUVEC and NIH 3T3 cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

Polycomb complex protein BMI-1 (UniProt: P35226; also known as Polycomb group RING finger protein 4, RING finger protein 51) is encoded by the BMI1 (also known as PCGF4, RNF51) gene (Gene ID: 100532731) in human. BMI-I (B cell-specific Moloney murine leukemia virus integration site 1) is a component of the polycomb repressive complex 1 (PRC1) that is required to maintain the transcriptionally repressive state of many genes, including Hox genes, throughout development. This complex acts via chromatin remodeling and modification of histones and mediates mono-ubiquitination of histone H2A lysine 119, rendering chromatin. The canonical PRC1 complex consists of four core subunits: CBX (polycomb; CBX2/4/6/7/8), PCGF (polycomb group factors; PCGF1 6), PHC (polyhomeotic homologues; PHC1/2/3), and RING E3 ligase (RING1A/B). BMI-I has three domains. The N-terminal RING domain, a central domain, and a C-terminal proline-rich domain. The RING domain forms a complex with RINGA/B proteins that constitutes the heterodimeric E3 ubiquitin ligase subunit of the PRC1 complex. The central is a ubiquitin-like (UBL) domain and is involved in protein-protein interactions and its binding partners include the polyhomeotic proteins (PHC1, PHC2, PHC3). The central domain of BMI-I is reported to be essential for its oncogenic activity and is shown to be essential for transcriptional repression activity. Higher expression of BMI1 gene is overexpressed in certain mantle cell lymphomas and several other cancers. This ZooMAb recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Gray, F., et al. (2016). Nat. Commun. 7; 13343).

Physical form

Purified recombinant mouse monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

Reconstitute lyophilized antibody pellet with 25μL of ultrapure water or Phosphate Buffered Saline (PBS). Please refer to our reconstitution protocol and the specific application guidance on the suggested starting dilutions and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service