Skip to Content
Merck
All Photos(1)

Key Documents

Z763144

Prism centrifuge adapters

for 0.5 mL tubes, pack of 8 ea

Synonym(s):

lab equipment accessory

Sign Into View Organizational & Contract Pricing

Select a Size

8 EA
€65.70

€65.70


Please contact Customer Service for Availability


Select a Size

Change View
8 EA
€65.70

About This Item

UNSPSC Code:
41103911
NACRES:
NB.78

€65.70


Please contact Customer Service for Availability

feature

Holds 0.5 mL tubes

packaging

pack of 8 ea

manufacturer/tradename

Labnet C1205-8

technique(s)

centrifugation: suitable

compatibility

for use with C1801
for use with Labnet Prism (TM) Mini Centrifuge

Looking for similar products? Visit Product Comparison Guide

Legal Information

Prism is a trademark of Labnet Intl., Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service