Yes, this product may only be used for volumes ranging from 1-10 mL.
Z757438
Sartorius Proline® Plus mechanical pippettor
single channel, volume 1-10 mL
Select a Size
€285.00
Select a Size
About This Item
€285.00
Recommended Products
feature
electronic: no
single-channel
packaging
pack of 1 ea
manufacturer/tradename
Sartorius 728090
volume
1-10 mL
General description
- Reduced pipetting forces prevent RSI and improve the results
- Accurate and precise
- Contamination free pipetting with Safe-Cone Filters
- Robust design
- Easiest pipette to maintain
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
Already Own This Product?
-
Please, is this pipette 1ml-10ml only? The picture doesn't represent that.
1 answer-
Helpful?
-