Z744246
Wilmad® Single Crystal Mounting Rod
size 4 mm, pack of 1
Synonym(s):
EPR Mounting Rod, Supasil Mounting Rod
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
packaging
pack of 1
manufacturer/tradename
Wilmad WG-857-Q
W × L
5 mm × 260 mm
size
4 mm
Looking for similar products? Visit Product Comparison Guide
General description
These rods are manufactured with the highest grade of synthetic quartz to eliminate E′ center defects and paramagnetic material contaminations.
Application
EPR
Legal Information
Wilmad is a registered trademark of SP Industries
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service