Z743685
Wash-N-Dry™ coverslip rack
XL, blue, holds 30 slipcover sizes including 18, 22 and 25 mm
Synonym(s):
slide rack
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
packaging
pack of 1 ea
manufacturer/tradename
Diversified Biotech WSDR-1100-B
color
blue
Looking for similar products? Visit Product Comparison Guide
General description
Holds 10 (30 for XL) Coverslips: Fast and easy cleaning, surface-treating and drying. Holds all slipcover sizes including 18, 22 and 25 mm.
Durable and Hassle Free: One piece polypropylene rack does not require assembly and is chemically and thermally resistant. Autoclavable.
Secure: Innovative design includes a notch to secure coverslips with a rubber band or loop of mono-filament.
Compatible: Fits inside a 100 mL beaker with space underneath for a small stir bar when submerged.
Note: The gap at the bottom of the Wash-N-Dry rack measures 8 mm (round coverslips also work with this product).
Durable and Hassle Free: One piece polypropylene rack does not require assembly and is chemically and thermally resistant. Autoclavable.
Secure: Innovative design includes a notch to secure coverslips with a rubber band or loop of mono-filament.
Compatible: Fits inside a 100 mL beaker with space underneath for a small stir bar when submerged.
Note: The gap at the bottom of the Wash-N-Dry rack measures 8 mm (round coverslips also work with this product).
Legal Information
Wash-N-Dry is a trademark of Diversified Biotech
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service