Key Documents
Z742664
Molecular Grinding Resin
with Pestle & Tubes
Select a Size
100 EA
€340.00
€340.00
Please contact Customer Service for Availability
Select a Size
Change View
100 EA
€340.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.74
€340.00
Please contact Customer Service for Availability
feature
with Pestle & Tubes
packaging
pkg of 100 ea
manufacturer/tradename
G-BioSciences 786-138PR
Related Categories
General description
Includes:
Molecular Grinding Resin 5 x 0.5 ml resin
Grinding Pestles (RNase Free) 100 x
Grinding Tubes (RNase Free) 100 x
Molecular Grinding Resin has been specifically developed for effectively grinding biological samples for extraction of DNA, RNA and proteins. The resin is high tensile micro-particles, which effectively disrupt nuclei and other cell organelles. Molecular Grinding Resin does not bind protein or nucleic acids. Simply mix the biological sample with Molecular Grinding Resin and grind or homogenize the sample. Unlike sand or other grinding materials, Molecular Grinding Resin is easy to work with and does not damage genomic DNA or high molecular mass proteins. We routinely use Molecular Grinding Resin for isolation of 100kb genomic DNA and RNA. Molecular Grinding Resin can be used with any homogenization technique, including high speed mechanical grinding such as Polytron and high-energy sonicators. Molecular Grinding Resin is also available with disposable matching pestles and tubes.
Legal Information
Molecular Grinding Resin is a trademark of Geno Technology, Inc.
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)