Skip to Content
Merck
All Photos(1)

Key Documents

Z742251

Lena Biosciences GradientEZ 3D Cell Migration System

6-lobe

Sign Into View Organizational & Contract Pricing

Select a Size

12 EA
€256.00

€256.00


Please contact Customer Service for Availability


Select a Size

Change View
12 EA
€256.00

About This Item

UNSPSC Code:
41122107
NACRES:
NB.22

€256.00


Please contact Customer Service for Availability

sterility

sterile

packaging

pack of 12 discs

manufacturer/tradename

Lena Biosciences SC-G002-0012

parameter

80 °C max. temp.

disc O.D. × I.D.

~16 mm × 11.5 mm

disc thickness

400 μm , dry state

color

white to off-white

pore size

90 % porosity

Looking for similar products? Visit Product Comparison Guide

General description

Chemical concentration gradients influence normal physiological processes and evolution and progression of many diseases.

GradientEZ substrates enable researchers to model gradient-dependent pathological conditions in 3D including cancer, inflammatory and autoimmune diseases, and to test therapeutic strategies in these models in vitro. You may use GradientEZ to apply, test and screen multiple test agents and their gradients in parallel, or sequentially with the in situ:
  • 3D cell cultures
  • 3D distributed population of invasive cells invading in 3D
  • 3D distributed suspension of cells taxing in 3D

These substrates build on proven SeedEZ compositions permitting generation of complex and reproducible gradients of soluble factors and substrate-bound molecules. All GradientEZ substrates support co- and counter-gradients of multiple test compounds applied in chemotaxis, invasion, chemo-invasion and angiogenesis assays.

Next, GradientEZ substrates enable concurrent testing of drug combination strategies in physiologically closer 3D models in vitro. This provides extraordinary knowledge base that cannot be elucidated in separate or sequential application of mono-strategies, with the added economic benefit of time, human resources and materials invested in research.

Other Notes

Fits into Petri dishes, 6-well to 12-well plates and 6-well inserts.
This substrate is not guaranteed to fit into a 24-well plate.

Legal Information

GradientEZ is a trademark of Lena Biosciences, Inc.
SeedEZ is a trademark of Lena Biosciences, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service