Skip to Content
Merck
All Photos(1)

Documents

Z742098

Western Blot Box

clear, size 8.9 cm × 6.5 cm × 2.5 cm, pk of 5

Synonym(s):

Western Blotting Box

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105300
NACRES:
NB.22

packaging

pk of 5

size

8.9 cm × 6.5 cm × 2.5 cm

color

clear

Looking for similar products? Visit Product Comparison Guide

General description

  • Perfect box for every blot
  • Reduce antibody usage
  • Hinged Lid minimizes evaporation
  • For halves, wholes, strips, or minis


No matter what size your Western blot is, there′s a box that fits it perfectly. With boxes that are correctly matched to common blot sizes, you can fit more blots on shakers/rockers while keeping reagent and antibody usage to an absolute minimum. Additionally, all corners are formed at 90° angles and the bottom surface is uniformly flat, further reducing consumption of costly reagents and antibodies (for example, popular mini-gels require only about 4mL).
At a price that makes them truly disposable, all sizes are engineered from crystal clear polystyrene. All sizes are also equipped with an easy-to-open hinged lid that reduces evaporation.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service