Skip to Content
Merck
All Photos(2)

Documents

Z720852

Nunc® Cell Factory, Nunclon® Δ surface

40-chamber CF, 25280 cm2, 8000 mL, 2/cs

Synonym(s):

cell culture bioreactor, cell culture chamber, cell factories

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000

material

polystyrene

sterility

sterile

packaging

pack of 1
case of 2

manufacturer/tradename

Nunc 139446

L × W

335mm × 205 mm

surface area

25280 cm2

working volume

8000 mL

color

clear

suitability

suitable for (industrial scale cell culture; production of biomaterials such as vaccines, monoclonal antibodies, etc.)

Looking for similar products? Visit Product Comparison Guide

General description

Nunc® Cell Factory, Nunclon® Δ surface

  • For large scale cell culture and production of biomaterials such as vaccines, monoclonal antibodies, etc
  • Ideal for adherent cells
  • Can be used for suspension culture as well
  • Growth kinetics unaltered from laboratory scale culture
  • Available in 1, 2, 4, 10 and 40 tray cell factory versions for easy scale-up
  • Compact design, large growth surface in limited space areas
  • Low risk of contamination
  • Easy to handle multi-layer cell factory stacks
  • Polystyrene trays assembled by sonic welding one on another
  • Not Autoclavable
  • Certified Nunclon® Δ surface treatment ensures excellent conditions for cell attachment and growth
  • Each unit is individually wrapped twice

Legal Information

Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Nunclon is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service