Key Documents
Z719706
Sorenson low binding standard tips
pack of 8 ea (Empty racks), non-sterile
Select a Size
10 EA
€34.20
€34.20
Please contact Customer Service for Availability
Select a Size
Change View
10 EA
€34.20
About This Item
UNSPSC Code:
41121600
NACRES:
NB.01
sterility:
non-sterile
packaging:
pack of 8 ea (Empty racks)
manufacturer/tradename:
Sorenson 28310
€34.20
Please contact Customer Service for Availability
material
colorless
polypropylene
sterility
non-sterile
packaging
pack of 8 ea (Empty racks)
manufacturer/tradename
Sorenson 28310
maximum volume
200 μL
Related Categories
General description
Tips feature a new surface technology designed for pipetting substances which tend to stick to the walls of pipette tips or for pipetting expensive reagents. Universal fit for research-grade pipettors.
Legal Information
Sorenson is a trademark of Sorenson BioScience, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)