Key Documents
Z719581
Sorenson low binding standard tips
maximum volume 200 μL, pkg of 96 tips CellRack, non-sterile
Select a Size
960 EA
€149.00
4800 EA
€564.00
€149.00
Please contact Customer Service for Availability
Select a Size
Change View
960 EA
€149.00
4800 EA
€564.00
About This Item
UNSPSC Code:
41121600
NACRES:
NB.01
volume:
200 μL
sterility:
non-sterile
feature:
barrier: no
packaging:
case of 4800 ea (50 racks of 96 tips)
pkg of 96 tips CellRack
pack of 960 ea (10 racks of 96 tips)
manufacturer/tradename:
Sorenson 15270T
€149.00
Please contact Customer Service for Availability
material
colorless
polypropylene
sterility
non-sterile
feature
barrier: no
packaging
case of 4800 ea (50 racks of 96 tips)
pkg of 96 tips CellRack
pack of 960 ea (10 racks of 96 tips)
manufacturer/tradename
Sorenson 15270T
maximum volume
200 μL
volume
200 μL
Related Categories
General description
Tips feature a new surface technology designed for pipetting substances which tend to stick to the walls of pipette tips or for pipetting expensive reagents. Universal fit for research-grade pipettors.
Legal Information
Sorenson is a trademark of Sorenson BioScience, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)