Skip to Content
Merck
All Photos(1)

Documents

Z660078

Bandelin TICKOPUR R 36 cleaning concentrate

special cleaner - tenside-free

Synonym(s):

detergent

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
47131800
NACRES:
NB.35

manufacturer/tradename

Bandelin

availability

available only in EU

General description

Special cleaner - tenside-free, non foaming, gentle to material, mildly alkaline, pH 9.9 (1 %), dosage 0.25 to 5 %, 1 to 10 min.

Contamination:

General contamination, oil, grease, distillation residues, organic and inorganic residues.

Objects to be cleaned:

Glass, optical glass, ceramics, plastics, rubber, steel, precious and light metals.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service