Key Documents
Z659886
Bandelin SONOREX Ultrasonic bath accessories
positioning lid for Z659843 and Z659045
Select a Size
1 EA
€99.00
€99.00
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
€99.00
About This Item
UNSPSC Code:
41103706
NACRES:
NB.74
€99.00
Please contact Customer Service for Availability
material
stainless steel
feature
for 4 beakers
packaging
pack of 1 ea
manufacturer/tradename
Bandelin 382
availability
not available in USA and Canada
General description
The insert strip is used to hold the insert containers. Insert cups are used when using chemically aggressive liquids, removing chemically aggressive or abrasive contamination or sonicating sample liquids (indirect sonication). Because nothing can be placed on the bottom of the ultrasonic bath, suitable perforated lids hold the insert cups. When using an insert strip, several insert cups with different liquids can be sonicated at the same time. In order to transfer the ultrasound to the insert vessel, the surface tension of the water must be reduced and therefore a cleaning solution must be used as a coupling liquid / contact liquid.
Features and Benefits
  • for holding insert cups
Legal Information
SONOREX is a trademark of Bandelin Electronic GmbH & Co. KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)