Key Documents
Z659347
Bandelin SONOPULS ultrasonic homogenizer accessories
processing vessel DG 4G
Synonym(s):
stainless steel sonification vessel
Select a Size
1 EA
€1,760.00
€1,760.00
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
€1,760.00
About This Item
UNSPSC Code:
41101520
NACRES:
NB.74
€1,760.00
Please contact Customer Service for Availability
material
stainless steel
packaging
pack of 1 ea
manufacturer/tradename
Bandelin 3608
availability
available only in EU
General description
Suitable for connection with a stepped standard horn with external thread (Z65,909-6 or Z65,911-8) with titanium flat tip TT13 (Z65,917-7).
Choose from one of the most recent versions:
Certificates of Analysis (COA)