Key Documents
Z659312
Bandelin SONOPULS ultrasonic homogenizer accessories
Sleeve adapter NA 29G for ground glass vessels
Synonym(s):
ultrasonic homogenizer adapter
Select a Size
1 EA
€153.00
€153.00
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
€153.00
About This Item
UNSPSC Code:
41101520
NACRES:
NB.74
€153.00
Please contact Customer Service for Availability
material
PTFE
packaging
pack of 1 ea
manufacturer/tradename
Bandelin 540
availability
available only in EU
General description
The standard ground joint adapter is used to create a tight connection between laboratory vessels with standard ground joints and SONOPULS ultrasonic homogenizers. This enables a wide range of sound irradiation setups and flow-through ultrasonic reactors to be realized. The adapter is screwed onto the external thread of the SONOPULS standard or booster horn.
Choose from one of the most recent versions:
Certificates of Analysis (COA)