Skip to Content
Merck
All Photos(1)

Key Documents

Z658014

Hettich® centrifuge buckets

for ROTOFIX 32A and UNIVERSAL 320/320R Centrifuges, bucket/carrier for rotor 1628

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€31.50

€31.50


Please contact Customer Service for Availability


Select a Size

Change View
1 EA
€31.50

About This Item

UNSPSC Code:
41103910
NACRES:
NB.22

€31.50


Please contact Customer Service for Availability

feature

Holds 1 x 15 mL tube

manufacturer/tradename

Hettich 1621

availability

available only in EU
not available in USA and Canada

technique(s)

centrifugation: suitable

compatibility

for use with rotor 1628 (Z665533)

Looking for similar products? Visit Product Comparison Guide

General description

  • compatible with rotor 1628 (Z665533)

Legal Information

Hettich is a registered trademark of Andreas Hettich GmbH & Co. KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service