Key Documents
Z640409
Eppendorf® epT.I.P.S. Reloads
volume range 0.5-20 μL
Synonym(s):
pipetting tips
About This Item
UNSPSC Code:
41121600
NACRES:
NB.01
L:
46 mm
maximum volume:
20 μL
volume range:
0.5-20 μL
sterility:
non-sterile
feature:
barrier: no
packaging:
pack of 960 (10 trays of 96 tips)
manufacturer/tradename:
Eppendorf® 0030 073.041
Eppendorf® 022491521
Product Z640409 is not currently sold in your country. Contact Technical Service
material
light gray tip
sterility
non-sterile
feature
barrier: no
packaging
pack of 960 (10 trays of 96 tips)
manufacturer/tradename
Eppendorf® 0030 073.041
Eppendorf® 022491521
L
46 mm
maximum volume
20 μL
volume range
0.5-20 μL
General description
Part of the ep T.I.P.S. box and reload system for contamination-free handling of refill trays, with plastic lids protecting the trays from contact during transfer to the working box.
Trays of tips which may be autoclaved in their packaging and stored clean for later use. Tips may be used straight from the reload tray, or loaded into ep T.I.P.S box (purchase separately)
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)