Skip to Content
Merck
All Photos(1)

Key Documents

Z637300

Mucocit-T instrument disinfecting detergent

volume 5 L

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
47131800

manufacturer/tradename

BRAND 44825

technique(s)

protein quantification: suitable

volume

5 L
5000 mL

Looking for similar products? Visit Product Comparison Guide

General description

Particularly suited for sensitive instruments. Free of phosphates, aldehydes, phenols and chlorine derivates. Fresh scent.

Microbiological efficiency / concentration for use:
  • Disinfection of instruments: Bacteria (incl. Tuberculosis and mycobacteria) and fungi with high organic burden acc. To DGHM/VAH directives: 1% (10 g/L)/ 1 hour., 2% (20 g/L)/ 30 min., 3% (30 g/L)
  • Limited virucidal efficacy (incl. HIV, HBV, HCV): 1% (10 g/L)/ 5 min.
  • Inactivation of human rotaviruses: 1% / 30 min., 2% / 5 min.
  • Inactivation of vaccinia viruses: 2% (20 g/L)/ 5 min.

Other Notes

BR44825 (India)

Legal Information

Mucocit is a trademark of Merz GmbH & Co. KG

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1 - Repr. 2 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service