Skip to Content
Merck
All Photos(1)

Key Documents

Z637297

Mucocit-T instrument disinfecting detergent

volume 2 L

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
47131800

manufacturer/tradename

BRAND 44822

volume

2 L
20000 mL

Looking for similar products? Visit Product Comparison Guide

General description

Particularly suited for sensitive instruments. Free of phosphates, aldehydes, phenols and chlorine derivates. Fresh scent.

Microbiological efficiency / concentration for use:
  • Disinfection of instruments: Bacteria (incl. Tuberculosis and mycobacteria) and fungi with high organic burden acc. To DGHM/VAH directives: 1% (10 g/L)/ 1 hour., 2% (20 g/L)/ 30 min., 3% (30 g/L)
  • Limited virucidal efficacy (incl. HIV, HBV, HCV): 1% (10 g/L)/ 5 min.
  • Inactivation of human rotaviruses: 1% / 30 min., 2% / 5 min.
  • Inactivation of vaccinia viruses: 2% (20 g/L)/ 5 min.

Other Notes

BR44822 (India)

Legal Information

Mucocit is a trademark of Merz GmbH & Co. KG

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service