Skip to Content
Merck
All Photos(1)

Key Documents

Z628026

BRAND® UV cuvettes

semi-micro, chamber volume 1.5-3 mL

Synonym(s):

10 mm, Cuvettes, UV Polymer, UV-Cuvettes

Sign Into View Organizational & Contract Pricing

Select a Size

100 EA
€39.70

€39.70


Please contact Customer Service for Availability


Select a Size

Change View
100 EA
€39.70

About This Item

UNSPSC Code:
41121813
NACRES:
SB.32

€39.70


Please contact Customer Service for Availability

feature

Disposable plastic
Wavelength range: 230 to 900 nm
semi-micro

manufacturer/tradename

BRAND 759150

chamber volume

1.5-3 mL

window W × H

4.5 mm × 2.3 mm

Looking for similar products? Visit Product Comparison Guide

General description

Ideally suited for concentration determinations through spectroscopy assays in water analysis, chemistry, and ife science applications. CERTIFIED LIFE SCIENCE QUALITY: Virgin raw materials, Manufactured under controlled room conditions, Intensive in-process controls, Visual inspections, Batch management, Final product inspection, Proven functionality.

Application

BRAND® UV cuvettes have been used in antigen adsorption and total protein content spectroscopy assays. [1]

Features and Benefits

  • Compatible with acid, alkaline solutions, and the majority of polar solvents
  • Low price with significantly lower risk of contamination as compared to other quartz glass cuvettes
  • Standard 10 mm light path.

Other Notes

BR759150 (India)

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

  1. Which document(s) contains shelf-life or expiration date information for a given product?

    If available for a given product, the recommended re-test date or the expiration date can be found on the Certificate of Analysis.

  2. How do I get lot-specific information or a Certificate of Analysis?

    The lot specific COA document can be found by entering the lot number above under the "Documents" section.

  3. How do I find price and availability?

    There are several ways to find pricing and availability for our products. Once you log onto our website, you will find the price and availability displayed on the product detail page. You can contact any of our Customer Sales and Service offices to receive a quote.  USA customers:  1-800-325-3010 or view local office numbers.

  4. What is the Department of Transportation shipping information for this product?

    Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product. 

  5. With what kind of plastic are the BRAND UV cuvettes made? Are they compatible with organic solvents like DMSO, as well as acids and bases?

    The BRAND (Brandtech Scientific) UV cuvettes are made of a proprietary plastic material that has excellent compatibility with acetone, acetonitrile, ethanol, isopropanol and DMSO (dimethyl sulfoxide), The material also has excellent compatibility with acids (hydrochloric acid, sulfuric acid and trichloroacetic acid solution), and bases (sodium hydroxide solution). The cuvettes are not compatible with hydrocarbons (pentane, hexane, heptane, benzene and toluene) or chlorinated solvents (chloroform, carbon tetrachloride and trichloroethylene).

  6. My question is not addressed here, how can I contact Technical Service for assistance?

    Ask a Scientist here.

Sven Jirschitzka et al.
Archiv der Pharmazie, 356(6), e2300072-e2300072 (2023-03-29)
Pertuzumab (Perjeta®) is a monoclonal antibody approved for the treatment of HER2-positive breast cancer. Before treatment, the concentrate must be diluted to obtain the ready-to-use infusion solution. Data on the storage stabilities of these preparations are lacking but important for all
Kyle Lakatos et al.
Human vaccines & immunotherapeutics, 16(8), 1957-1968 (2020-01-30)
More effective rotavirus vaccines are essential for preventing extensive diarrheal morbidity and mortality in children under five years of age in low-resource regions. Nonreplicating rotavirus vaccines (NRRV) administered parenterally provide an alternate vaccination method to the current licensed oral vaccine.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service