Skip to Content
Merck
All Photos(1)

Key Documents

Z617792

Greiner centrifuge tubes with screw cap

capacity 12 mL, size × diam. × L × 16.8 mm × 100 mm, conical bottom, skirt

Synonym(s):

centrifuge tubes, culture tubes, plastic centrifuge tube, plastic culture tube, screw cap centrifuge tube

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121703
NACRES:
NB.11
Product Z617792 is not currently sold in your country. Contact Technical Service

material

colorless polystyrene tube
conical bottom
screw top blue polyethylene cap

sterility

sterile

feature

skirt

packaging

case of 1000

manufacturer/tradename

Greiner 164161

capacity

12 mL

size diam. × L

× 16.8 mm × 100 mm

Looking for similar products? Visit Product Comparison Guide

General description

Polystyrene tubes with closures are ideally suited for optical measurements due to its high clarity. Available with screw or bayonetc ap in blue, black or white and choice of skirted on non-skirted.

Application

Greiner centrifuge tubes with screw cap is suitable for processing, storage, and centrifugation of samples.

Features and Benefits

  • High clarity as the tube is made of polystyrene
  • Cap is made of high-density polyethylene
  • RNase, DNase, human DNA, and pyrogens free
  • Non-autoclavable

Legal Information


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service