Skip to Content
Merck
All Photos(1)

Key Documents

Z614459

Vivaspin 6 centrifugal concentrator

MWCO 5 kDa

Synonym(s):

Spin Concentrator, Vivaspin 6

Sign Into View Organizational & Contract Pricing

Select a Size

100 EA
€797.00

€797.00


Please contact Customer Service for Availability


Select a Size

Change View
100 EA
€797.00

About This Item

UNSPSC Code:
41104916
NACRES:
NB.24

€797.00


Please contact Customer Service for Availability

material

polycarbonate housing
polyethersulfone membrane

description

PES membrane

sterility

non-sterile

feature

holdup volume 10 μL

packaging

package of 100 ea

manufacturer/tradename

Sartorius VS0612

parameter

10000 × g max. centrifugal force

dead stop volume

30 μL

membrane area

2.5 cm2

tube size

~15 mL

working volume

6 mL

pore size

5 kDa MWCO

Looking for similar products? Visit Product Comparison Guide

General description

The Vivaspin 6 centrifugal concentrator has been developed to offer increased volume flexibility and performance. Vivaspin 6 can be used in either swing bucket or fixed angle rotors (carriers accepting 15 mL and 50 mL bottles). Vivaspin features twin vertical membranes for unparalleled filtration speeds and 100X plus concentrations. Remaining volume is easy to read off the printed scales on the side of the concentrator and the modified dead stop pocket further simplifies direct pipette recovery of the final concentrate.

  • Vivaspin centrifugal concentrators offer the optimal solution to any concentration or buffer exchange application with their broad range of volume capacities and MWCOs.
  • Highest flow rates and recoveries are achieved due to their particular vertical membrane design.
  • Incomparably safe sample handling and convenience is assured by the patented dead-stop technology allowing direct recovery of the concentrate without further re-spin.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service