Z614262
Vivaspin 2 centrifugal concentrators
MWCO 50 kDa
Select a Size
€192.00
Select a Size
About This Item
€192.00
Recommended Products
material
polycarbonate housing
polyethersulfone membrane
sterility
non-sterile
feature
holdup volume 10 μL
packaging
package of 25 ea
manufacturer/tradename
Sartorius VS0231
parameter
8000 × g max. centrifugal force
dead stop volume
8 μL
membrane area
1.2 cm2
tube size
~15 mL
working volume
2 mL
pore size
50 kDa MWCO
General description
- Vivaspin centrifugal concentrators offer the optimal solution to any concentration or buffer exchange application with their broad range of volume capacities and MWCOs.
- Highest flow rates and recoveries are achieved due to their particular vertical membrane design.
- Incomparably safe sample handling and convenience is assured by the patented dead-stop technology allowing direct recovery of the concentrate without further re-spin.
Choose from one of the most recent versions:
Certificates of Analysis (COA)