Z382493
SigmaScreen™ Poly-D-lysine coated plates
96-well, clear
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41100000
Recommended Products
material
clear plastic plate
flat bottom clear wells
description
flat bottom
sterility
sterile
feature
lid
manufacturer/tradename
Sigma-Aldrich
size
96 wells
color
clear
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
General description
SigmaScreen Poly D-Lysine (PDL) coated multiwell plates are coated with a synthetic PDL molecular weight range of 70-150 kDa. By coating the negatively charged polystyrene surface with PDL, the resulting positively charged surface has been shown to improve cell survival under low serum or serum-free conditions. SigmaScreen PDL coated multiwell plates can be used with a variety of cell lines including HEK-293 and mouse 3T3. For a more comprehensive listing of cell lines used with PDL coated multiwell plates, please refer to the PDL coated plate technical bulletin. Manufactured under ISO9002 in Sigma′s GMP facility, Sigma′s PDL coated multiwell plates are bioburden tested (fungal and bacterial) and endotoxin free.
Legal Information
SigmaScreen is a trademark of Sigma-Aldrich Co. LLC
Storage Class Code
12 - Non Combustible Liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Shengjun Chang et al.
Neuroscience letters, 441(1), 134-138 (2008-07-04)
HIF-1 is believed to play a critical role in hypoxia/ischemia (H/I) preconditioning protection in neonatal brain. Recently, it has been shown that hydrogen peroxide (H(2)O(2)) may contribute to H/I preconditioning in rat primary neurons. We hypothesize that H(2)O(2) produced during
Jian-Fu Chen et al.
The Journal of cell biology, 190(5), 867-879 (2010-09-08)
Skeletal muscle satellite cells are adult stem cells responsible for postnatal skeletal muscle growth and regeneration. Paired-box transcription factor Pax7 plays a central role in satellite cell survival, self-renewal, and proliferation. However, how Pax7 is regulated during the transition from
Gudrun Strauss et al.
The Journal of experimental medicine, 206(6), 1379-1393 (2009-06-03)
CD95 is a multifunctional receptor that induces cell death or proliferation depending on the signal, cell type, and cellular context. Here, we describe a thus far unknown function of CD95 as a silencer of T cell activation. Naive human T
Zhanyan Fu et al.
Molecular and cellular neurosciences, 42(1), 45-55 (2009-05-26)
Neuroligins (NLGs) are postsynaptic cell adhesion molecules that are thought to function in synaptogenesis. To investigate the role of NLGs on synaptic transmission once the synapse is formed, we transfected neuroligin-2 (NLG-2) in cultured mouse cerebellar granule cells (CGCs), and
Corentin Laulier et al.
Nucleic acids research, 39(14), 5935-5944 (2011-04-05)
Homology-directed repair (HDR) is essential to limit mutagenesis, chromosomal instability (CIN) and tumorigenesis. We have characterized the consequences of HDR deficiency on anaphase, using markers for incomplete chromosome separation: DAPI-bridges and Ultra-fine bridges (UFBs). We show that multiple HDR factors
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service