Key Documents
Z374962
PCR microtube and cap strips
capacity 0.2 mL
Select a Size
1 PKG
€320.00
€320.00
Please contact Customer Service for Availability
Select a Size
Change View
1 PKG
€320.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.22
€320.00
Please contact Customer Service for Availability
material
cap (strips)
colorless tube
sterility
sterile
feature
neck thread push fit
packaging
pkg of 250 strips (8 tubes or caps per strip)
technique(s)
PCR: suitable
capacity
0.2 mL
working volume
0.2 mL
binding type
non-treated surface
General description
Strips of eight tubes connected with double bridges to avoid accidental separation. Caps also are in strips of eight. Can be cut apart to use individually if desired.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Protocols
Hot Start dNTPs block DNA polymerase until heat activation, enhancing PCR specificity.
Method for amplification of DNA from damaged DNA sources. Particularly useful for DNA extracted from old samples.
MTP™ Taq DNA Polymerase is a recombinant thermostable enzyme from Thermus aquaticus expressed in E. coli and purified using a proprietary process to minimize levels of contaminating DNA.
Reactions using REDTaq® DNA polymerase are formulated as any PCR mixtures. There are no additional reaction preparation steps or protocol changes required.
See All