Key Documents
Z332364
Bottle adapters for BRAND® Dispensers and Burettes
GL 33 thread, GL 40 bottle, ETFE
Select a Size
1 EA
€19.00
€19.00
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
€19.00
About This Item
UNSPSC Code:
41121815
NACRES:
NB.01
€19.00
Please contact Customer Service for Availability
material
ETFE
manufacturer/tradename
BRAND 704390
fitting
GL 40 bottle
GL 33 thread
Related Categories
General description
For all Dispensette® models as well as seripettor® and QuikSip
Other Notes
BR704390 (India)
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Dispensette is a registered trademark of BRAND GMBH + CO KG
QuikSip is a trademark of BRAND GMBH + CO KG
seripettor is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)