Key Documents
Z325015
BRAND® BLAUBRAND® bulb pipette, calibrated to deliver (TD, EX)
one mark, capacity 9 mL
Select a Size
6 EA
€62.40
€62.40
Please contact Customer Service for Availability
Select a Size
Change View
6 EA
€62.40
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
€62.40
Please contact Customer Service for Availability
material
AR-Glas®
description
total delivery Ex + 15 sec waiting time
feature
class AS
packaging
pack of 6 ea
manufacturer/tradename
BRAND 29711
capacity
9 mL
Related Categories
General description
Additional to DIN
All BLAUBRAND bulb pipettes are supplied with a batch number and an accompanying batch certificate in the original packaging.
According to DIN EN ISO 648.
Other Notes
BR29711 (India)
Legal Information
AR-Glas is a registered trademark of Schott AG
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Protocols
This page contains detailed information on working with and handling pipettes and pipetting aids.