Skip to Content
Merck
All Photos(1)

Key Documents

Z0377

Sigma-Aldrich

Anti-Zyxin antibody, Mouse monoclonal

clone ZOL301, purified from hybridoma cell culture

Synonym(s):

Anti-ESP-2, Anti-HED-2

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
€601.00

€601.00


Please contact Customer Service for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1408

Request a Bulk Order

Select a Size

Change View
200 μL
€601.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

€601.00


Please contact Customer Service for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1408

Request a Bulk Order

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

ZOL301, monoclonal

form

buffered aqueous solution

mol wt

antigen 82-84 kDa

species reactivity

mouse, rat, human

packaging

antibody small pack of 25 μL

technique(s)

immunocytochemistry: suitable
indirect ELISA: suitable
microarray: suitable
western blot: 1-2 μg/mL using total cell extract of HeLa cells.

isotype

IgG2a

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ZYX(7791)
mouse ... Zyx(22793)
rat ... Zyx(114636)

General description

Anti-Zyxin antibody, Mouse monoclonal (mouse IgG2a isotype) is derived from the hybridoma ZOL301 produced by the fusion of mouse myeloma cells (NS1 cells) and splenocytes from BALB/c mice immunized with a synthetic peptide corresponding to human Zyxin conjugated to KLH via an N-terminal added cysteine. Zyxin is a relatively low abundance component of focal adhesion plaques, stress fibers, and dynamic cell membrane areas such as the leading edge. It may also be found in the cell nucleus. Zyxin is a member of the LIM domain protein family that also includes LPP (Lipoma preferred partner), Trip6 (thyroid hormone interacting protein 6), and Ajuba. It is a phosphoprotein and comprises two major domains, an extended N-terminal domain that contains proline-rich sequence, and a C-terminal domain that contains a functional nuclear export signal and three double zinc-finger cysteine and histidine-rich LIM domains.

Application

Anti-Zyxin antibody, Mouse has been used in:
  • Western blotting
  • Immunofluorescence staining[1]
  • Enzyme linked immunosorbent assay (ELISA)
  • Immunocytochemistry

Biochem/physiol Actions

Zyxin interacts with multiple cytoskeletal, signaling and other proteins such as α -actinin, vasodilator-stimulated phosphoprotein (VASP), Mena, LIM-only cysteine-rich proteins (CRP1, CRP2, CRP3/MLP), Vav, breast cancer anti-estrogen resistance protein 1 (p130Cas), CRK-associated substrate-related protein/ (Human Enhancer of Filamentation 1) (CasL/HEF1, and serine-arginine(SR)-related protein (SON) DNA-binding protein. During mitosis a fraction of zyxin was reported to associate with the tumor suppressor h-warts/LATS1 (large tumor suppressor kinase 1) on the mitotic apparatus. Zyxin has been implicated in the regulation and modulation of actin filaments organization, cell adhesion, cell motility, mitosis, and signal transduction. Zyxin may shuttle between focal adhesion plaques and the nucleus. Involvement of zyxin in gene transcription by its association with transcription factors in the nucleus has been suggested.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anna Cykowska et al.
Frontiers in molecular medicine, 3, 1050487-1050487 (2023-04-20)
Malignant melanoma is the most lethal form of skin cancer. Y-box binding protein 1 (YB-1) plays a prominent role in mediating metastatic behavior by promoting epithelial-to-mesenchymal transition (EMT). Migratory melanoma cells exhibit two major migration modes: elongated mesenchymal or rounded
Zyxin and paxillin proteins: focal adhesion plaque LIM domain proteins go nuclear
Wang Y and Gilmore TD
Biochimica et Biophysica Acta - Molecular Cell Research, 1593(2-3), 115-120 (2003)
Rezvan Rostampour et al.
Health science reports, 7(8), e2288-e2288 (2024-08-05)
Breast cancer (BC) is considered one of the most common malignant tumors leading to death in women, and genetic factors have a crucial role in BC pathogenesis. Zyxin (ZYX) is one of these factors that may be important in p53
Genetic ablation of zyxin causes Mena/VASP mislocalization, increased motility, and deficits in actin remodeling
Hoffman LM, et al.
The Journal of Cell Biology, 172(5), 771-782 (2006)
Zyxin, a regulator of actin filament assembly, targets the mitotic apparatus by interacting with h-warts/LATS1 tumor suppressor
Hirota T, et al.
The Journal of Cell Biology, 149(5), 1073-1086 (2000)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service