Skip to Content
Merck
All Photos(1)

Documents

X3629

Sigma-Aldrich

X80

≥98% (HPLC)

Synonym(s):

5-(5-{[1-(3-carboxyphenyl)-3-methyl-5-oxo-1,5-dihydro-4H-pyrazol-4-ylidene]methyl}-2-furyl)-2-chlorobenzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H15ClN2O6
CAS Number:
Molecular Weight:
450.83
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

orange

solubility

DMSO: >40 mg/mL

storage temp.

2-8°C

SMILES string

CC1=NN(c2cccc(c2)C(O)=O)C(=O)\C1=C/c3ccc(o3)-c4ccc(Cl)c(c4)C(O)=O

InChI

1S/C23H15ClN2O6/c1-12-17(21(27)26(25-12)15-4-2-3-14(9-15)22(28)29)11-16-6-8-20(32-16)13-5-7-19(24)18(10-13)23(30)31/h2-11H,1H3,(H,28,29)(H,30,31)/b17-11-

InChI key

XZRBYOCWWJMJRZ-BOPFTXTBSA-N

Application

X80 has been used as an inhibitor of nucleotide excision repair (NER) pathway to study its effects on 8-hydroxy-2′-deoxyguanosine (8-OHdG) excision in the zygote post-fertilization.

Biochem/physiol Actions

X80 is an inhibitor of xeroderma pigmentosum group A protein (XPA protein) required for the removal of all types of DNA lesions repaired by nucleotide excision repair Including both transcription coupled and global genomic nucleotide excision repair.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiao-Bo Dong et al.
Journal of agricultural and food chemistry, 67(4), 1261-1268 (2019-01-10)
Many organisms possess the ability to produce metal-binding proteins to absorb cadmium. Metallothioneins, an important family of cysteine-rich metal-binding proteins, have been isolated and well characterized. However, Lentinula edodes may have a different type of cadmium-binding protein that contains fewer
Qian Wu et al.
Journal of agricultural and food chemistry, 65(1), 216-223 (2016-12-16)
Anthocyanins were isolated from blueberry wine lees using Sephadex LH-20 column chromatography and semipreparative high-performance liquid chromatography (semipreparative HPLC) and then identified by HPLC-DAD-ESI-MS/MS. Our results show that malvidin-3-hexose (Mv-3-hex) and malvidin-3-(6'acetyl)-hexose (Mv-3-ace-hex) are the major components in the anthocyanin
Wendy G Cárdenas-Coronel et al.
Journal of agricultural and food chemistry, 64(1), 302-309 (2015-12-03)
Quality and compositional changes were determined in noni fruit harvested at five ripening stages, from dark-green to thaslucent-grayish. Fruit ripening was accompanied by acidity and soluble solids accumulation but pH diminution, whereas the softening profile presented three differential steps named
Yan-Yu Cui et al.
BMC biotechnology, 21(1), 5-5 (2021-01-11)
At present, microorganism has been considered as important factors that threaten to buried pipelines with disbonded coatings. Aiming at the problem of unknown corrosion mechanism of sulfate-reducing bacteria (SRB), a series of studies have been carried out in this paper.
Fertilization stimulates 8-hydroxy-2?-deoxyguanosine repair and antioxidant activity to prevent mutagenesis in the embryo
Lord T and Aitken RJ
Developmental Biology, 406(1), 1-13 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service