Skip to Content
Merck
All Photos(1)

Key Documents

WH0164832M1

Sigma-Aldrich

Monoclonal Anti-LONRF2 antibody produced in mouse

clone 1C7, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-FLJ45273, Anti-LON peptidase N-terminal domain and ring finger 2, Anti-MGC126711, Anti-MGC126713, Anti-RNF192

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€489.00

€489.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
€489.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€489.00


Please contact Customer Service for Availability

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1C7, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable

isotype

IgG2aκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LONRF2(164832)

General description

The gene encoding LON peptidase N-terminal domain and ring finger 2 (LONRF2) is localized on human chromosome 2q11.2.

Immunogen

LONRF2 (NP_940863, 1 a.a. ~ 108 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MKNYEQALQDASAACQNEPLLIKGHQVKAQALSGLGRSKEVLKEFLYCLALNPECNSVKKEAQKVMCEVLFSATANVHENLTSSIQSRLKAQGHSHMNAQALLEEGDA

Biochem/physiol Actions

LON peptidase N-terminal domain and ring finger 2 (LONRF2) has been linked to auto-immune diseases like rhematoid arthritis.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

CLARITY: C hi L dhood A rthritis R isk Factor I dentification ST ud Y. Investigating the role of aberrant DNA methylation in juvenile idiopathic arthritis
J.A. Ellis
Pediatrics null
Sabyasachi Senapati et al.
European journal of human genetics : EJHG, 23(4), 530-535 (2014-07-24)
Studies in European populations have contributed to a better understanding of the genetics of complex diseases, for example, in coeliac disease (CeD), studies of over 23 000 European samples have reported association to the HLA locus and another 39 loci. However

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service