Skip to Content
Merck
All Photos(4)

Key Documents

WH0054751M10

Sigma-Aldrich

Monoclonal Anti-FBLIM1 antibody produced in mouse

clone 5E11, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-CAL, Anti-DKFZp434G171, Anti-FBLP1, Anti-RP11169K16.5, Anti-filamin binding LIM protein 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€489.00

€489.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
€489.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€489.00


Please contact Customer Service for Availability

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

5E11, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FBLIM1(54751)

General description

This gene encodes a protein with an N-terminal filamin-binding domain, a central proline-rich domain, and, multiple C-terminal LIM domains. This protein localizes at cell junctions and may link cell adhesion structures to the actin cytoskeleton. This protein may be involved in the assembly and stabilization of actin-filaments and likely plays a role in modulating cell adhesion, cell morphology and cell motility. This protein also localizes to the nucleus and may affect cardiomyocyte differentiation after binding with the CSX/NKX2-5 transcription factor. Alternative splicing results in multiple transcript variants encoding different isoforms. (provided by RefSeq)

Immunogen

FBLIM1 (NP_060026, 270 a.a. ~ 373 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
VTCARCIGDESFALGSQNEVYCLDDFYRKFAPVCSICENPIIPRDGKDAFKIECMGRNFHENCYRCEDCRILLSVEPTDQGCYPLNNHLFCKPCHVKRSAAGCC

Biochem/physiol Actions

FBLIM1 (filamin binding LIM protein 1) plays an important role in association between the cell membrane and the actin cytoskeleton. It mainly interacts with MIG-2 (mitogen-inducible gene 2 protein), filamin and VASP (vasodilator-stimulated phosphoprotein), thus controlling cell shape and movements. In cardiomyocytes, it interacts with transcription factor CSX/NKX2-5 (cardiac-specific homeobox) and enhances the differentiation. The FBLIM1 gene is upregulated in osteoarthritis chondrocytes and might be involved with osteoarthritis pathogenesis. In various carcinoma cells, the FBLIM1-associated signaling in disrupted which leads to abnormal Src activation and anoikis resistance in cells. This gene is downregulated in breast cancer cells. In esophageal cancer cells, it suppresses invasion partly by enhancing degradation of β-catenin. However, in glioma cells FBLIM1 promotes migration as well as invasion.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Migfilin's elimination from osteoarthritic chondrocytes further promotes the osteoarthritic phenotype via ?-catenin upregulation.
Gkretsi V, et al.
Biochemical and Biophysical Research Communications, 430, 494-499 (2013)
Migfilin interacts with Src and contributes to cell-matrix adhesion-mediated survival signaling.
Zhao J, et al.
The Journal of Biological Chemistry, 284, 34308-34320 (2009)
Migfilin, a-parvin and ?-parvin are differentially expressed in ovarian serous carcinoma effusions, primary tumors and solid metastases.
Davidson B, et al.
Gynecologic Oncology, 128, 364-370 (2013)
Mitogen-inducible Gene-2 (MIG2) and migfilin expression is reduced in samples of human breast cancer.
Gkretsi V, et al.
Anticancer Research, 33, 1977-1981 (2013)
Migfilin regulates esophageal cancer cell motility through promoting GSK-3?-mediated degradation of ?-catenin.
He H, et al.
Molecular Cancer Research, 10, 273-281 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service