Skip to Content
Merck
All Photos(8)

Key Documents

WH0009097M4

Sigma-Aldrich

Monoclonal Anti-USP14 antibody produced in mouse

clone 6D6, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-TGT, Anti-ubiquitin specific peptidase 14 (tRNA-guanine transglycosylase)

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€489.00

€489.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
€489.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€489.00


Please contact Customer Service for Availability

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6D6, monoclonal

form

buffered aqueous solution

species reactivity

rat, mouse, human

technique(s)

indirect ELISA: suitable
indirect immunofluorescence: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... USP14(9097)

Related Categories

General description

Ubiquitin specific peptidase 14 (USP14) is a 494 amino acid protein containing a 45kDa catalytic domain at its amino-terminus. Its ubiquitin-like (Ubl) domain associates with 19S regulatory particle (RP) 26S proteasome. This binding activates the deubiquitinating activity of the protein. USP14 is part of the ubiquitin-specific processing (UBP) family of proteases.

Immunogen

USP14 (NP_005142, 395 a.a. ~ 494 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
PQKEVKYEPFSFADDIGSNNCGYYDLQAVLTHQGRSSSSGHYVSWVKRKQDEWIKFDDDKVSIVTPEDILRLSGGGDWHIAYVLLYGPRRVEIMEEESEQ

Biochem/physiol Actions

Ubiquitin specific peptidase 14 (USP14) releases ubiquitin from the proteasome-targeted ubiquitinated proteins resulting in regeneration of ubiquitin at the proteasome. It is involved in the degradation of chemokine receptor CXCR4. Under endoplasmic reticulum (ER) stress, USp14 binds to ER stress receptor, which is involved in unfolded protein response (UPR), a mechanism to control the accumulation of unfolded proteins within the ER lumen. Its overexpression leads to ER-associated degradation (ERAD) pathway inhibition.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structure and mechanisms of the proteasome-associated deubiquitinating enzyme USP14
The Embo Journal (2005)
Deubiquitination of CXCR4 by USP14 is critical for both CXCL12-induced CXCR4 degradation and chemotaxis but not ERK activation
Marjelo A Mines
The Journal of Biological Chemistry (2008)
USP14 inhibits ER-associated degradation via interaction with IRE1alpha
Atsushi Nagai
Biochemical and Biophysical Research Communications (2009)
Zhanyu Ding et al.
Molecular cell, 73(6), 1150-1161 (2019-02-23)
The 26S proteasome is the ATP-dependent protease responsible for regulating the proteome of eukaryotic cells through degradation of mainly ubiquitin-tagged substrates. In order to understand how proteasome responds to ubiquitin signal, we resolved an ensemble of cryo-EM structures of proteasome
Jayashree Chadchankar et al.
PloS one, 14(11), e0225145-e0225145 (2019-11-09)
USP14 is a cysteine protease deubiquitinase associated with the proteasome and plays important catalytic and allosteric roles in proteasomal degradation. USP14 inhibition has been considered a therapeutic strategy for accelerating degradation of aggregation-prone proteins in neurodegenerative diseases and for inhibiting

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service