Skip to Content
Merck
All Photos(2)

Key Documents

WH0008452M1

Sigma-Aldrich

Monoclonal Anti-CUL3 antibody produced in mouse

clone 1A3, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-cullin 3

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1A3, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CUL3(8452)

General description

CUL3 is a component of a ubiquitin E3 ligase that is essential for mitotic division (Sumara et al., 2007 [PubMed 17543862]).[supplied by OMIM

Immunogen

CUL3 (AAH39598, 301 a.a. ~ 400 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
KLFSRVPNGLKTMCECMSSYLREQGKALVSEEGEGKNPVDYIQGLLDLKSRFDRFLLESFNNDRLFKQTIAGDFEYFLNLNSRSPEYLSLFIDDKLKKGV

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sayaka Yoshida et al.
Clinical and experimental nephrology, 22(6), 1251-1257 (2018-06-06)
Pseudohypoaldosteronism type II (PHAII) is a hereditary hypertensive disease caused by mutations in four genes: WNK1, WNK4, Kelch-like3 (KLHL3), and cullin3 (CUL3). Recently, it was revealed that CUL3-KLHL3 E3 ligase complex ubiquitinates WNK1 and WNK4, leading to their degradation, and
Chien-Ming Lin et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 33(1), 1051-1061 (2018-08-28)
The Kelch-like 3 ( KLHL3) mutations contributed to the most common causative genes in patients with pseudohypoaldosteronism type II (PHAII); however, the molecular mechanisms of PHAII-causing mutations in BTB domain of KLHL3 in vivo have not been investigated. We generated
Sutton Mooney et al.
Cells, 8(4) (2019-04-13)
The turnip Brassica rapa has important economic value and represents a good model system to study gene function in crop plants. ERF/AP2 transcription factors are a major group of proteins that are often involved in regulating stress-responses and developmental programs.
Zhi Chen et al.
Journal of cellular physiology, 234(11), 19977-19989 (2019-04-05)
Inflammation is one of the major causes of intervertebral disc degeneration (IDD). Emerging evidence has revealed that increase in the levels of pro-inflammatory cytokines, such as interleukin 6 (IL-6) and tumor necrosis factor alpha (TNF-α), can activate a variety of
Mitchell A Pallett et al.
Journal of virology, 93(10) (2019-03-01)
Viral infection of cells is sensed by pathogen recognition receptors that trigger an antiviral innate immune response, and consequently viruses have evolved countermeasures. Vaccinia virus (VACV) evades the host immune response by expressing scores of immunomodulatory proteins. One family of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service