Skip to Content
Merck
All Photos(2)

Key Documents

WH0006274M1

Sigma-Aldrich

Monoclonal Anti-S100A3 antibody produced in mouse

clone 1D4, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-S100 calcium binding protein A3, Anti-S100E

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€416.00

€416.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
€416.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€416.00


Please contact Customer Service for Availability

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1D4, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... S100A3(6274)

General description

The protein encoded by this gene is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21. This protein has the highest content of cysteines of all S100 proteins, has a high affinity for Zinc, and is highly expressed in human hair cuticle. The precise function of this protein is unknown. (provided by RefSeq)

Immunogen

S100A3 (NP_002951, 1 a.a. ~ 101 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MARPLEQAVAAIVCTFQEYAGRCGDKYKLCQAELKELLQKELATWTPTEFRECDYNKFMSVLDTNKDCEVDFVEYVRSLACLCLYCHEYFKDCPSEPPCSQ

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ran Tao et al.
Experimental and therapeutic medicine, 13(6), 2812-2818 (2017-06-08)
The fifth most common cancer worldwide is hepatocellular carcinoma (HCC), which has an annual mortality rate of ~800,000. Although surgical procedures for HCC, such as hepatic resection and liver transplantation, have progressed and the outcomes of patients have improved, HCC

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service