Skip to Content
Merck
All Photos(4)

Key Documents

WH0005026M1

Sigma-Aldrich

Monoclonal Anti-P2RX5 antibody produced in mouse

clone 1C5, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-MGC47755, Anti-P2X5, Anti-P2X5R, Anti-purinergic receptor P2X, ligand-gated ion channel, 5

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€489.00

€489.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
€489.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€489.00


Please contact Customer Service for Availability

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1C5, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... P2RX5(5026)

General description

The product of this gene belongs to the family of purinoceptors for ATP. This receptor functions as a ligand-gated ion channel. Several characteristic motifs of ATP-gated channels are present in its primary structure, but, unlike other members of the purinoceptors family, this receptor has only a single transmembrane domain. Three transcript variants encoding distinct isoforms have been identified for this gene. (provided by RefSeq)

Immunogen

P2RX5 (NP_002552, 126 a.a. ~ 224 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
DGACSKDSDCHAGEAVTAGNGVKTGRCLRRENLARGTCEIFAWCPLETSSRPEEPFLKEAEDFTIFIKNHIRFPKFNFSKSNVMDVKDRSFLKSCHFGP

Biochem/physiol Actions

P2RX5 (Purinergic receptor P2X, ligand-gated ion channel, 5) functions as a ATP-gated ion channel. In humans, it is predicted to form trimeric structure with another member of this family, P2X1. Upregulated P2RX5 expression has been observed during CD4+ T cell activation. It has been studied that activated P2RX5 plays an important role in the T cell polarity and immunoregulation, when it is recruited to the surface of the activated CD4+ T cells.[1]

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Smita Kotnis et al.
Molecular pharmacology, 77(6), 953-960 (2010-03-13)
P2X5 is a member of the P2X family of ATP-gated nonselective cation channels, which exist as trimeric assemblies. P2X5 is believed to trimerize with another member of this family, P2X1. We investigated the single-nucleotide polymorphism (SNP) at the 3' splice
Pierre Abramowski et al.
PloS one, 9(9), e104692-e104692 (2014-09-03)
Members of the P2X family of ligand-gated cation channels (P2RX) are expressed by various cell types including neurons, smooth- and cardiac muscle cells, and leukocytes. The channels mediate signalling in response to extracellular ATP. Seven subunit isoforms (P2RX1-P2RX7) have been

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service