Skip to Content
Merck
All Photos(5)

Key Documents

WH0004760M1

Sigma-Aldrich

Monoclonal Anti-NEUROD1 antibody produced in mouse

clone 3H8, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-BETA2, Anti-BHF1, Anti-NEUROD, Anti-neurogenic differentiation 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€489.00

€489.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
€489.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

€489.00


Please contact Customer Service for Availability

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3H8, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
indirect ELISA: suitable
indirect immunofluorescence: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NEUROD1(4760)

General description

Neuronal differentiation 1 (NeuroD1) belongs to the family of a basic helix-loop-helix transcription factor. The NEUROD1 gene is localized on human chromosome 2q31.3 and is expressed in developing neurons.

Immunogen

NEUROD1 (AAH09046, 201 a.a. ~ 300 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
QDMPPHLPTASASFPVHPYSYQSPGLPSPPYGTMDSSHVFHVKPPPHAYSAALEPFFESPLTDCTSPSFDGPLSPPLSINGNFSFKHEPSAEFEKNYAFT

Biochem/physiol Actions

Neuronal differentiation 1 (NeuroD1) may mediate the genesis of functional neurons from glial cells. Its higher expression favors neural differentiation in elder rats. NeuroD1 may be crucial for reversing functional impairment by favoring axonal regeneration in nerve injury. Various mutations in NEUROD1 reported have direct implications in the pathophysiology of early-onset diabetes, retinal dystrophy, nonsyndromic retinitis pigmentosa, and neurological abnormalities.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lack of association between NeuroD1/D6 gene polymorphism and heroin dependence in Han-chinese male population
Tsou CC, et al.
Journal of Medical Sciences null
Muhua Lai et al.
Experimental neurology, 327, 113215-113215 (2020-01-29)
Neurogenic differentiation 1 (NeuroD1) is mainlyexpressed in developing neurons where it plays critical roles in neuronal maturation and neurite elongation. The potential role and mechanism of NeuroD1 in adult axonal regeneration is not clear. The present study used synapsin (SYN)
Feng Wang et al.
Investigative ophthalmology & visual science, 56(1), 150-155 (2014-12-06)
Mutations in the same gene can lead to different clinical phenotypes. In this study, we aim to identify novel genotype-phenotype correlations and novel disease genes by analyzing an unsolved autosomal recessive retinitis pigmentosa (ARRP) Han Chinese family. Whole exome sequencing
Abhijeet Pataskar et al.
The EMBO journal, 35(1), 24-45 (2015-11-01)
Cell fate specification relies on the action of critical transcription factors that become available at distinct stages of embryonic development. One such factor is NeuroD1, which is essential for eliciting the neuronal development program and possesses the ability to reprogram

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service