Skip to Content
Merck
All Photos(3)

Key Documents

WH0001289M1

Sigma-Aldrich

Monoclonal Anti-COL5A1 antibody produced in mouse

clone 2F4, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-collagen, type V, alpha 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€489.00

€489.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
€489.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€489.00


Please contact Customer Service for Availability

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2F4, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... COL5A1(1289)

General description

The collagen type V α1 chain (COL5A1) gene is mapped to human chromosome 9q34.3. The gene codes for a minor fibrillar collagen called type V collagen.
This gene encodes an alpha chain for one of the low abundance fibrillar collagens. Fibrillar collagen molecules are trimers that can be composed of one or more types of alpha chains. Type V collagen is found in tissues containing type I collagen and appears to regulate the assembly of heterotypic fibers composed of both type I and type V collagen. This gene product is closely related to type XI collagen and it is possible that the collagen chains of types V and XI constitute a single collagen type with tissue-specific chain combinations. Mutations in this gene are associated with Ehlers-Danlos syndrome, types I and II. (provided by RefSeq)

Immunogen

COL5A1 (AAH08760, 1 a.a. ~ 591 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MGPPGPPGPRGPSGAPGADGPQGPPGGIGNPGAVGEKGEPGEAGEPGLPGEGGPPGPKGERGEKGESGPSGAAGPPGPKGPPGDDGPKGSPGPVGFPGDPGPPGEPGPAGQDGPPGDKGDDGEPGQTGSPGPTGEPGPSGPPGKRGPPGPAGPEGRQGEKGAKGEAGLEGPPGKTGPIGPQGAPGKPGPDGLRGIPGPVGEQGLPGSPGPDGPPGPMGPPGLPGLKGDSGPKGEKGHPGLIGLIGPPGEQGEKGDRGLPGPQGSSGPKGEQGITGPSGPIGPPGPPGLPGPPGPKGAKGSSGPTGPKGEAGHPGPPGPPGPPGEVIQPLPIQASRTRRNIDASQLLDDGNGENYVDYADGMEEIFGSLNSLKLEIEQMKRPLGTQQNPARTCKDLQLCHPDFPDGEYWVDPNQGCSRDSFKVYCNFTAGGSTCVFPDKKSEGARITSWPKENPGSWFSEFKRGKLLSYVDAEGNPVGVVQMTFLRLLSASAHQNVTYHCYQSVAWQDAATGSYDKALRFLGSNDEEMSYDNNPYIRALVDGCATKKGYQKTVLEIDTPKVEQVPIVDIMFNDFGEASQKFGFEVGPACFMG

Biochem/physiol Actions

Collagen type Vα1 chain (COL5A1) intercalates with type I collagen and forms collagen fibrils. Mutations in the gene claims to affect mechanical properties of collagenous tissues and lead to musculoskeletal diseases. In addition, polymorphism in the gene is also associated with the development of vascular Ehlers-Danlos syndrome (EDS), which is characterized by fragile, hyperextensible skin and joint laxity.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Familial Ehlers-Danlos syndrome with lethal arterial events caused by a mutation in COL5A1.
Monroe GR, et al.
American Journal of Medical Genetics. Part A, 167, 1196-1203 (2015)
Mapping of the human COL5A1 gene to chromosome 9q34.3
Caridi G, et al.
Human Genetics, 90, 174-176 (1992)
Eric A Kirk et al.
Experimental physiology, 101(12), 1581-1592 (2016-11-04)
What is the central question of the study? Do COL5A1 gene variants, previously reported to have diminished transcript stability, manifest in physiological phenotypes of quadriceps muscle-tendon contractile properties and mechanical stiffness in humans? What is the main finding and its
Julide Altinisik et al.
The American journal of sports medicine, 43(7), 1784-1789 (2015-04-22)
Tennis elbow entails pain and tenderness over the lateral epicondyle. The exact cause of the condition is not fully understood. Type V collagen is a minor fibrillar collagen that intercalates with type I collagen and forms collagen fibrils. It is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service