Skip to Content
Merck
All Photos(6)

Key Documents

WH0000475M1

Sigma-Aldrich

Monoclonal Anti-ATOX1 antibody produced in mouse

clone 2E6, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-ATX1, Anti-ATX1 antioxidant protein 1 homolog (yeast), Anti-HAH1, Anti-MGC138453, Anti-MGC138455

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€489.00

€489.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
€489.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€489.00


Please contact Customer Service for Availability

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2E6, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
indirect immunofluorescence: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ATOX1(475)

General description

This gene encodes a copper chaperone that plays a role in copper homeostasis by binding and transporting cytosolic copper to ATPase proteins in the trans-Golgi network for later incorporation to the ceruloplasmin. This protein also functions as an antioxidant against superoxide and hydrogen peroxide, and therefore, may play a significant role in cancer carcinogenesis. Because of its cytogenetic location, this gene represents a candidate gene for 5q-syndrome. (provided by RefSeq)
ATOX1 (antioxidant 1 copper chaperone) is a metallochaperone which is also called as HAH1. It is a small cytosolic protein. ATOX1 is located on human chromosome 5q33.

Immunogen

ATOX1 (NP_004036, 1 a.a. ~ 68 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MPKHEFSVDMTCGGCAEAVSRVLNKLGGVKYDIDLPNKKVCIESEHSMDTLLATLKKTGKTVSYLGLE

Application

Monoclonal Anti-ATOX1 antibody has been used in immunoblot analysis.[1]

Biochem/physiol Actions

ATOX1 (antioxidant 1 copper chaperone) participates in the human copper modulation system. It plays a major role in the migration of breast cancer cells. ATOX1 helps in the transportation of copper to the cell secretory pathway.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ye-Jin Kim et al.
Metallomics : integrated biometal science, 11(8), 1430-1440 (2019-07-19)
Copper (Cu) is a tightly regulated micronutrient that functions as a structural or catalytic cofactor for specific proteins essential for a diverse array of biological processes. While the study of the extremely rare genetic diseases, Menkes and Wilson, has highlighted
Copper chaperone Atox1 plays role in breast cancer cell migration.
Blockhuys S and Wittung-Stafshede P
Biochemical and Biophysical Research Communications, 483(1), 301-304 (2017)
Metallochaperone Atox1 transfers copper to the NH2-terminal domain of the Wilson's disease protein and regulates its catalytic activity.
Walker JM, et al.
The Journal of Biological Chemistry, 277(31), 27953-27959 (2002)
The structural flexibility of the human copper chaperone Atox1: Insights from combined pulsed EPR studies and computations.
Levy AR, et al.
Protein Science, 26(8), 1609-1618 (2017)
ATOX1 gene silencing increases susceptibility to anticancer therapy based on copper ionophores or chelating drugs.
Barresi V, et al.
Journal of Inorganic Biochemistry, 156, 145-152 (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service