Skip to Content
Merck
All Photos(2)

Key Documents

VIV404002

SynVivo SynTox 3D Toxicology model starter kit, IMN2 radial configuration

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
€2,120.00

€2,120.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EACH
€2,120.00

About This Item

UNSPSC Code:
60104506
NACRES:
NA.84

€2,120.00


Please contact Customer Service for Availability

packaging

case of 1 ea

manufacturer/tradename

SynVivo 404002

storage temp.

room temp

General description

10 - IMN2 SynTox 2 μm slit barrier chips, 100 ft Tygon Tubing, 25 pack - Slide Clamps, 50 pack - 1 mL Syringe with Luer-Lok Tip, 50 pack - 24 Gauge Needle x 0.5 in long, SynVivo Pneumatic Primer, 2 - Five Stopcock Manifold 4-Way all FLL

SynTox 3D toxicology model chip- Barrier type: Slits, Slit width: 2 μm, Distance between slits: 50 μm, Travel/barrier width: 50 μm, Outer Channel width: 200 μm, Device depth: 100 μm. This chip is used to create the SynTox model in monoculture, co or tri-culture configurations.

SynTox is the only commercially available 3D toxicology model with real-time optical monitoring and multi-compartment, multi-cellular architecture and low reagent requirements. Other benefits of this platform are:

  • Physiologically realistic morphological, fluidic and 3D cellular conditions
  • Universal platform with architecture specific of desired organ
  • Significant reduction in cost and time
  • Robust and easy to use protocols
  • Compatible with standard analytical instruments for both on chip and off chip assays including omic methodologies for systems biology and bioinformatic analysis

Linkage

LInk to Publications
application text

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service