Skip to Content
Merck
All Photos(1)

Key Documents

V9881

Sigma-Aldrich

Vitronectin from bovine plasma

lyophilized powder, BioReagent, suitable for cell culture

Synonym(s):

Serum spreading factor

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.75
Product V9881 is not currently sold in your country. Contact Technical Service

biological source

bovine plasma

Quality Level

product line

BioReagent

form

lyophilized powder

packaging

pkg of 50 μg

technique(s)

cell culture | mammalian: suitable

surface coverage

0.1 μg/cm2

solubility

water: soluble 1.0 mL, clear, colorless

UniProt accession no.

shipped in

ambient

storage temp.

2-8°C

Gene Information

bovine ... VTN(507525)

Application

Vitronectin can be used with cells with integrin receptors that bind vitronectin, including platelets, endothelial cells, melanoma cells, and osteosarcoma. It is recommended as a cell culture substratum at 0.1 μg/cm2. Optimal conditions for attachment depend on cell line and application.

Biochem/physiol Actions

Vitronectin binds to glycosaminoglycans, is incorporated as an inhibitor to the membrane cytolytic attack complex of the complement system, has interaction with thrombin and antithrombin III during coagulation, and may play a role in the coagulation pathway.

Components

Vitronectin is a glycoprotein present in plasma and tissues, and is one of the major cell adhesion proteins, along with fibronectin, in plasma. The N-terminal residues of vitronectin are identical to somatomedin B and are followed by an R-G-D sequence that interacts with a specific cell-surface receptor. The central domain of vitronectin is enriched with hydrophobic residues. Closer to the C-terminus, it has an arginine rich region that allows for heparin binding activity.

Caution

The lyophilized product should be stored desiccated at 2-8°C. Under these conditions it is stable for at least 2 years. Aliquots should be stored at -20°C or -70°C, but repeated freezing and thawing should be avoided.

Preparation Note

This product is supplied as a cell culture tested lyophilized powder from buffered saline. It is recommended to use polypropylene vessels during handling, due to the strong affinity of vitronectin for hydrophilic surfaces. Reconstitution of this product can be done with 1 mL of water per vial. The stock solution should then be filter-sterilized using a 0.2μm membrane and aliquoted and frozen for long term storage.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stephanie L Gupton et al.
The Journal of cell biology, 198(4), 657-676 (2012-08-22)
Mena is an Ena/VASP family actin regulator with roles in cell migration, chemotaxis, cell-cell adhesion, tumor cell invasion, and metastasis. Although enriched in focal adhesions, Mena has no established function within these structures. We find that Mena forms an adhesion-regulated
Yusuke Arima et al.
Acta biomaterialia, 26, 72-81 (2015-08-27)
We examined the effect of surface chemistry on adsorption of fibronectin (Fn) and vitronectin (Vn) and subsequent cell adhesion, employing self-assembled monolayers (SAMs) of alkanethiols carrying terminal methyl (CH3), hydroxyl groups (OH), carboxylic acid (COOH), and amine (NH2). More Fn
Joseph Mathew Kalappurakkal et al.
Cell, 177(7), 1738-1756 (2019-05-21)
Glycosylphosphatidylinositol-anchored proteins (GPI-APs) are a major class of lipid-anchored plasma membrane proteins. GPI-APs form nanoclusters generated by cortical acto-myosin activity. While our understanding of the physical principles governing this process is emerging, the molecular machinery and functional relevance of GPI-AP

Articles

3D cell culture overview. Learn about 2D vs 3D cell culture, advantages of 3D cell culture, and techniques available to develop 3D cell models

Cancer stem cell media, spheroid plates and cancer stem cell markers to culture and characterize CSC populations.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service